ENA390346999 Enamine

1-(4-fluorophenyl)-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD17189515
  • Cas Number : 1245149-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125300

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125300
Availability details for
Quantity
  • Availability
  • Add to Cart