ENA390347020 Enamine

2-(1,5-dimethyl-1H-pyrazol-3-yl)acetonitrile

  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1,5-dimethyl-1H-pyrazol-3-yl)acetonitrile
Inchi Code 1S/C7H9N3/c1-6-5-7(3-4-8)9-10(6)2/h5H,3H2,1-2H3
Inchi Key VATJIQKCMWMOIF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-125348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125348
Availability details for
Quantity
  • Availability
  • Add to Cart