ENA390347022 Enamine

6-amino-1-propyl-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD11987151
  • Cas Number : 1153515-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-125350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125350
Availability details for
Quantity
  • Availability
  • Add to Cart