ENA390347023 Enamine

1-bromo-2-(bromomethyl)-3-fluorobenzene

  • MDL number MFCD10698675
  • Cas Number : 1548-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125351
Availability details for
Quantity
  • Availability
  • Add to Cart