ENA390347024 Enamine

3H-imidazo[4,5-b]pyridine-7-carboxylic acid

  • MDL number MFCD05723323
  • Cas Number : 78316-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122856

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122856
Availability details for
Quantity
  • Availability
  • Add to Cart