ENA390347025 Enamine

3-methyl-5-oxo-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD03011581
  • Cas Number : 79932-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125353
Availability details for
Quantity
  • Availability
  • Add to Cart