ENA390347030 Enamine

(4-chloronaphthalen-1-yl)boronic acid

  • MDL number MFCD18411295
  • Cas Number : 147102-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125358

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125358
Availability details for
Quantity
  • Availability
  • Add to Cart