ENA390347037 Enamine

methyl 2-(methylamino)-2-(pyridin-2-yl)propanoate

  • MDL number MFCD12150922
  • Cas Number : 1184002-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125366
Availability details for
Quantity
  • Availability
  • Add to Cart