ENA390347046 Enamine

5-(2H-1,2,3,4-tetrazol-5-yl)pyridin-2-amine

  • MDL number MFCD18258940
  • Cas Number : 13566-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 308-309
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125377
Availability details for
Quantity
  • Availability
  • Add to Cart