ENA390347047 Enamine

2-(chloromethyl)-5-(propan-2-yl)-1,3,4-thiadiazole

  • MDL number MFCD18250343
  • Cas Number : 1260670-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106789

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106789
Availability details for
Quantity
  • Availability
  • Add to Cart