ENA390347057 Enamine

2-ethylquinoline-4,6-diamine

  • MDL number MFCD13192505
  • Cas Number : 1092352-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125423
Availability details for
Quantity
  • Availability
  • Add to Cart