ENA390347070 Enamine

1-(4-fluorophenyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD11202307
  • Cas Number : 106461-31-8
  • Molecular Weight : 227.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-125438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125438
Availability details for
Quantity
  • Availability
  • Add to Cart