ENA390347071 Enamine

3-[4-fluoro-3-(trifluoromethyl)phenyl]-3-oxopropanenitrile

  • MDL number MFCD16083874
  • Cas Number : 677713-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125441

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125441
Availability details for
Quantity
  • Availability
  • Add to Cart