ENA390347074 Enamine

5-amino-4,4-dimethylpentan-1-ol

  • MDL number MFCD19374424
  • Cas Number : 740784-18-3
  • Molecular Weight : 131.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-125447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125447
Availability details for
Quantity
  • Availability
  • Add to Cart