ENA390347083 Enamine

3-bromo-N1-methylbenzene-1,2-diamine

  • MDL number MFCD12024390
  • Cas Number : 1150617-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123858
Availability details for
Quantity
  • Availability
  • Add to Cart