ENA390347090 Enamine

8-(iodomethyl)quinoline

  • MDL number MFCD06410700
  • Cas Number : 215606-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125523

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125523
Availability details for
Quantity
  • Availability
  • Add to Cart