ENA390347095 Enamine

1-{4-[(4-fluorophenyl)sulfanyl]phenyl}methanamine

  • MDL number MFCD12191936
  • Cas Number : 1156220-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125529
Availability details for
Quantity
  • Availability
  • Add to Cart