ENA390347098 Enamine

4-chloro-6-methoxypyridine-3-carbonitrile

  • MDL number MFCD18415526
  • Cas Number : 1261493-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125532
Availability details for
Quantity
  • Availability
  • Add to Cart