ENA390347100 Enamine

6-bromo-5-methylpyridine-2-carbonitrile

  • MDL number MFCD11044272
  • Cas Number : 450844-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125534
Availability details for
Quantity
  • Availability
  • Add to Cart