ENA390347103 Enamine

9-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD09732823
  • Cas Number : 51991-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205...207
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125538
Availability details for
Quantity
  • Availability
  • Add to Cart