ENA390347108 Enamine

ethyl 4-cyclopropyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD12924385
  • Cas Number : 936720-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125544
Availability details for
Quantity
  • Availability
  • Add to Cart