ENA390347109 Enamine

2,2-dimethyl-N-[2-(5-sulfamoylthiophen-2-yl)ethyl]propanamide

  • MDL number MFCD11101759
  • Cas Number : 1152603-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125547

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125547
Availability details for
Quantity
  • Availability
  • Add to Cart