ENA390347121 Enamine

2-amino-5-nitropyridine-3-carboxylic acid

  • MDL number MFCD03427671
  • Cas Number : 6760-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 318-319
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125584
Availability details for
Quantity
  • Availability
  • Add to Cart