ENA390347123 Enamine

1-nonyl-1H-indole

  • MDL number MFCD19529729
  • Cas Number : 1406828-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125586
Availability details for
Quantity
  • Availability
  • Add to Cart