ENA390347130 Enamine

3-(3-chloro-4-fluorophenyl)-3-oxopropanenitrile

  • MDL number MFCD02260787
  • Cas Number : 267880-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124434

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124434
Availability details for
Quantity
  • Availability
  • Add to Cart