ENA390347132 Enamine

4-(difluoromethoxy)-1-phenyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11523855
  • Cas Number : 1152603-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125595
Availability details for
Quantity
  • Availability
  • Add to Cart