ENA390347137 Enamine

3-(trifluoromethyl)-1,2,4-oxadiazol-5-amine

  • MDL number MFCD20697766
  • Cas Number : 35488-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-105
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125600
Availability details for
Quantity
  • Availability
  • Add to Cart