ENA390347140 Enamine

(5-methyl-1,3,4-thiadiazol-2-yl)methanol

  • MDL number MFCD18432908
  • Cas Number : 869108-51-0
  • Molecular Weight : 130.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-methyl-1,3,4-thiadiazol-2-yl)methanol
Inchi Code 1S/C4H6N2OS/c1-3-5-6-4(2-7)8-3/h7H,2H2,1H3
Inchi Key LZCLPLGMVCURFT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-125603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125603
Availability details for
Quantity
  • Availability
  • Add to Cart