ENA390347144 Enamine

3-oxo-2H,3H,5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid

  • MDL number MFCD22396895
  • Cas Number : 1432680-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125608

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-125608
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart