ENA390347156 Enamine

3-ethoxycyclobutan-1-one

  • MDL number MFCD21755693
  • Cas Number : 30830-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125675
Availability details for
Quantity
  • Availability
  • Add to Cart