ENA390347162 Enamine

methyl 2-(6-chloropyridin-3-yl)acetate

  • MDL number MFCD15526721
  • Cas Number : 717106-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125687
Availability details for
Quantity
  • Availability
  • Add to Cart