ENA390347163 Enamine

(chloromethoxy)benzene

  • MDL number MFCD00623650

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125688
Availability details for
Quantity
  • Availability
  • Add to Cart