ENA390347166 Enamine

3,4-dihydro-2H-1-benzopyran-6-sulfonic acid

  • MDL number MFCD21963234
  • Cas Number : 127461-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125702
Availability details for
Quantity
  • Availability
  • Add to Cart