ENA390347173 Enamine

(4-chlorophenyl)[4-(trifluoromethyl)phenyl]methanamine

  • MDL number MFCD12621309
  • Cas Number : 1019353-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125709
Availability details for
Quantity
  • Availability
  • Add to Cart