ENA390347179 Enamine

N-[4-(2-aminoethoxy)phenyl]methanesulfonamide hydrochloride

  • MDL number MFCD20259717
  • Cas Number : 522602-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125715

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125715
Availability details for
Quantity
  • Availability
  • Add to Cart