ENA390347182 Enamine

3,5-dichloro-4-(2-methoxyethoxy)aniline

  • MDL number MFCD11647684
  • Cas Number : 1098376-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58...60
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125719

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125719
Availability details for
Quantity
  • Availability
  • Add to Cart