ENA390347183 Enamine

2-({1-[(tert-butoxy)carbonyl]piperidin-3-yl}methoxy)acetic acid

  • MDL number MFCD09753925
  • Cas Number : 231622-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125720

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125720
Availability details for
Quantity
  • Availability
  • Add to Cart