ENA390347192 Enamine

4-cyclopropyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD16707047
  • Cas Number : 1247103-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125758
Availability details for
Quantity
  • Availability
  • Add to Cart