ENA390347193 Enamine

4-(phenoxymethyl)aniline

  • MDL number MFCD06804490
  • Cas Number : 57181-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125759

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-125759
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart