ENA390347196 Enamine

4-chloro-3-cyanobenzoic acid

  • MDL number MFCD09999808
  • Cas Number : 117738-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124638

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-124638
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart