ENA390347199 Enamine

N-methyl-4-(2-nitrophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD17258259
  • Cas Number : 1007563-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125766

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125766
Availability details for
Quantity
  • Availability
  • Add to Cart