ENA390347217 Enamine

2-amino-4-methanesulfonyl-N-methyl-N-(oxan-4-yl)butanamide hydrochloride

  • MDL number MFCD23701891
  • Cas Number : 1427890-35-4
  • Molecular Weight : 314.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-N-methyl-4-(methylsulfonyl)-N-(tetrahydro-2H-pyran-4-yl)butanamide hydrochloride
Inchi Code 1S/C11H22N2O4S.ClH/c1-13(9-3-6-17-7-4-9)11(14)10(12)5-8-18(2,15)16;/h9-10H,3-8,12H2,1-2H3;1H
Inchi Key SGMKXHPSKMZMIR-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-125788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125788
Availability details for
Quantity
  • Availability
  • Add to Cart