ENA390347220 Enamine

N-(2,2,2-trifluoroethyl)-5,6,7,8-tetrahydroquinolin-8-amine

  • MDL number MFCD21790636
  • Cas Number : 1432679-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125795
Availability details for
Quantity
  • Availability
  • Add to Cart