ENA390347225 Enamine

benzyl(dodecyl)amine

  • MDL number MFCD00093619
  • Cas Number : 1687-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124772

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124772
Availability details for
Quantity
  • Availability
  • Add to Cart