ENA390347227 Enamine

4-[(dimethylamino)methyl]-2-nitroaniline

  • MDL number MFCD16689337
  • Cas Number : 1249667-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125829
Availability details for
Quantity
  • Availability
  • Add to Cart