ENA390347230 Enamine

4-(methylsulfanyl)pentanoic acid

  • MDL number MFCD16693341
  • Cas Number : 75974-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125832

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125832
Availability details for
Quantity
  • Availability
  • Add to Cart