ENA390347231 Enamine

2-methoxy-1-(3,4,5-trifluorophenyl)ethan-1-amine

  • MDL number MFCD18657905
  • Cas Number : 1270344-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125833
Availability details for
Quantity
  • Availability
  • Add to Cart