ENA390347235 Enamine

3-{[(pyrimidin-2-yl)amino]methyl}benzoic acid

  • MDL number MFCD13166802
  • Cas Number : 1179835-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125837
Availability details for
Quantity
  • Availability
  • Add to Cart