ENA390347237 Enamine

methyl 4-isocyanatobutanoate

  • MDL number MFCD03411283
  • Cas Number : 27678-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125843

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125843
Availability details for
Quantity
  • Availability
  • Add to Cart