ENA390347240 Enamine

2-[chloro(phenyl)methyl]-5-(3-methylbutyl)-1,3,4-oxadiazole

  • MDL number MFCD11546046
  • Cas Number : 1097053-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125847

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125847
Availability details for
Quantity
  • Availability
  • Add to Cart